5-[(Methylamino)methyl]-3-phenylisoxazole 97%
|
Peligro
|
APOSOR14866-1GEA
0
EUR
APOSOR14866-1G
APOSOR14866-250MG
5-[(Methylamino)methyl]-3-phenylisoxazole 97%
5-[(Methylamino)methyl]-3-phenylisoxazole
Formula:
C₁₁H₁₂N₂O MW: 188,226 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09055377 Núm. CAS: 852431-00-6 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...