1-(2-Aminopropyl)-1H-imidazole dihydrochloride 95%

APOSOR12978-1GEA 0 EUR
APOSOR12978-1G APOSOR12978-250MG
1-(2-Aminopropyl)-1H-imidazole dihydrochloride 95%
1-(2-Aminopropyl)-1H-imidazole dihydrochloride
Formula: C₆H₁₁N₃·2ClH
MW: 198,094 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD05861371
Núm. CAS: 1158449-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR