3-(4-methylphenoxy)benzylamine hydrochloride

APOSOR12861-250MGEA 0 EUR
APOSOR12861-250MG APOSOR12861-1G
3-(4-methylphenoxy)benzylamine hydrochloride
3-(4-methylphenoxy)benzylamine hydrochloride
Formula: C₁₄H₁₅NO·ClH
MW: 249,736 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD03840100
Núm. CAS: 154108-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR