2-[(Piperidin-4-yl)oxy]pyrazine dihydrochloride
Proveedor: Apollo Scientific
|
Atención
|
APOSOR12809-50MGEA
40.8
EUR
APOSOR12809-50MG
2-[(Piperidin-4-yl)oxy]pyrazine dihydrochloride
2-[(Piperidin-4-yl)oxy]pyrazine dihydrochloride
Formula:
C₉H₁₃N₃O·2ClH MW: 252,141 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD03840122 Núm. CAS: 950649-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...