2-(piperidin-4-yloxy)pyrimidine dihydrochloride
APOSOR12808-1GEA
0
EUR
APOSOR12808-1G
APOSOR12808-5G
2-(piperidin-4-yloxy)pyrimidine dihydrochloride
2-(piperidin-4-yloxy)pyrimidine dihydrochloride
Formula:
C₉H₁₃N₃O·2ClH MW: 252,141 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD03840123 Núm. CAS: 950649-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...