6-(3-Bromophenyl)-2-chloronicotinic acid
APOSOR12507-1GEA
0
EUR
APOSOR12507-1G
6-(3-Bromophenyl)-2-chloronicotinic acid
6-(3-Bromophenyl)-2-chloronicotinic acid
Formula:
C₁₂H₇BrClNO₂ MW: 312,546 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09027894 Núm. CAS: 946384-94-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...