3,4-Dihydro-4-methyl-7-[(methylamino)methyl]-2H-pyrido[3,2-b][1,4]oxazine 97%
APOSOR12415-1GEA
0
EUR
APOSOR12415-1G
APOSOR12415-250MG
3,4-Dihydro-4-methyl-7-[(methylamino)methyl]-2H-pyrido[3,2-b][1,4]oxazine 97%
3,4-Dihydro-4-methyl-7-[(methylamino)methyl]-2H-pyrido[3,2-b][1,4]oxazine
Formula:
C₁₀H₁₅N₃O MW: 193,246 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09817509 Núm. CAS: 921938-85-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...