5-(3,5-Dichlorophenoxy)-2-furoyl chloride
APOSOR1241-250MGEA
0
EUR
APOSOR1241-250MG
APOSOR1241-1G
5-(3,5-Dichlorophenoxy)-2-furoyl chloride
5-(3,5-Dichlorophenoxy)-2-furoyl chloride
Formula:
C₁₁H₅Cl₃O₃ MW: 291,515 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00173779 Núm. CAS: 175277-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...