6-Chloro-N-cyclopropyl-2-(methylsulphanyl)pyrimidin-4-amine
Proveedor: Apollo Scientific
APOSOR11613-5GEA
408
EUR
APOSOR11613-5G
APOSOR11613-1G
6-Chloro-N-cyclopropyl-2-(methylsulphanyl)pyrimidin-4-amine
6-Chloro-N-cyclopropyl-2-(methylsulphanyl)pyrimidin-4-amine
Formula:
C₈H₁₀ClN₃S MW: 215,703 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09800982 Núm. CAS: 951884-05-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...