4-Chloro-6-(pentylamino)pyrimidine 98%
APOSOR11508-5GEA
0
EUR
APOSOR11508-5G
APOSOR11508-1G
4-Chloro-6-(pentylamino)pyrimidine 98%
4-Chloro-6-(pentylamino)pyrimidine
Formula:
C₉H₁₄ClN₃ MW: 199,681 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD09800956 Núm. CAS: 941294-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...