Ethyl (2-acetyl-5-chlorophenoxy)acetate
Proveedor: Apollo Scientific
APOSOR111601-1GEA
187
EUR
APOSOR111601-1G
Ethyl (2-acetyl-5-chlorophenoxy)acetate
Ethyl (2-acetyl-5-chlorophenoxy)acetate
Formula:
C₁₂H₁₃ClO₄ MW: 256,682 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD27949053 Núm. CAS: 30335-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...