Ethyl 5-bromo-3-methyl-1-benzofuran-2-carboxylate
Proveedor: Apollo Scientific
APOSOR111595-1GEA
187
EUR
APOSOR111595-1G
Ethyl 5-bromo-3-methyl-1-benzofuran-2-carboxylate
Ethyl 5-bromo-3-methyl-1-benzofuran-2-carboxylate
Formula:
C₁₂H₁₁BrO₃ MW: 283,118 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD03965818 Núm. CAS: 150612-59-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...