Methyl 3-(4-nitrophenyl)sulphanylpropanoate

APOSOR111556-1GEA 0 EUR
APOSOR111556-1G
Methyl 3-(4-nitrophenyl)sulphanylpropanoate
Methyl 3-(4-nitrophenyl)sulphanylpropanoate
Formula: C₁₀H₁₁NO₄S
MW: 241,264 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD12169615
Núm. CAS: 7597-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR