Methyl 1-(4-formylphenyl)-1H-pyrazole-3-carboxylate

Proveedor: Apollo Scientific

APOSOR111324-1GEA 0 EUR
APOSOR111324-1G
Methyl 1-(4-formylphenyl)-1H-pyrazole-3-carboxylate
Methyl 1-(4-formylphenyl)-1H-pyrazole-3-carboxylate
Formula: C₁₂H₁₀N₂O₃
MW: 230,219 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD29918601
Núm. CAS: 2108305-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR