[4-(5-Oxo-1,5-dihydro-4H-1,2,4-triazol-4-yl)phenyl]acetic acid
Proveedor: Apollo Scientific
APOSOR111285-1GEA
891
EUR
APOSOR111285-1G
APOSOR111285-50MG
APOSOR111285-500MG
APOSOR111285-250MG
[4-(5-Oxo-1,5-dihydro-4H-1,2,4-triazol-4-yl)phenyl]acetic acid
[4-(5-Oxo-1,5-dihydro-4H-1,2,4-triazol-4-yl)phenyl]acetic acid
Formula:
C₁₀H₉N₃O₃ MW: 219,197 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD30529700 Núm. CAS: 1695360-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...