Methyl 2-[4-(3,4-dimethoxyphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]propanoate
Proveedor: Apollo Scientific
APOSOR111256-1GEA
564
EUR
APOSOR111256-1G
Methyl 2-[4-(3,4-dimethoxyphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]propanoate
Methyl 2-[4-(3,4-dimethoxyphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]propanoate
Formula:
C₁₄H₁₇N₃O₅ MW: 307,302 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD30481136 Núm. CAS: 2197054-06-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...