2-Azepan-1-ylnicotinonitrile
Proveedor: Apollo Scientific
APOSOR111236-1GEA
0
EUR
APOSOR111236-1G
2-Azepan-1-ylnicotinonitrile
2-Azepan-1-ylnicotinonitrile
Formula:
C₁₂H₁₅N₃ MW: 201,268 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD08144877 Núm. CAS: 684648-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...