Ethyl 4-imidazo[1,2-a]pyridin-2-ylbenzoate
Proveedor: Apollo Scientific
APOSOR111165-1GEA
0
EUR
APOSOR111165-1G
Ethyl 4-imidazo[1,2-a]pyridin-2-ylbenzoate
Ethyl 4-imidazo[1,2-a]pyridin-2-ylbenzoate
Formula:
C₁₆H₁₄N₂O₂ MW: 266,295 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD22573876 Núm. CAS: 175153-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...