Ethyl 2-naphthylacetate

APOSOR111125-5GEA 0 EUR
APOSOR111125-5G APOSOR111125-1G
Ethyl 2-naphthylacetate
Ethyl 2-naphthylacetate
Formula: C₁₄H₁₄O₂
MW: 214,26 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00014323
Núm. CAS: 2876-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR