Ethyl 1-(5-methyl-6-phenylthieno[2,3-d]pyrimidin-4-yl)piperidine-4-carboxylate
Proveedor: Apollo Scientific
APOSOR111064-1GEA
326
EUR
APOSOR111064-1G
Ethyl 1-(5-methyl-6-phenylthieno[2,3-d]pyrimidin-4-yl)piperidine-4-carboxylate
Ethyl 1-(5-methyl-6-phenylthieno[2,3-d]pyrimidin-4-yl)piperidine-4-carboxylate
Formula:
C₂₁H₂₃N₃O₂S MW: 381,491 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD28148252 Núm. CAS: 1858257-30-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...