Hard hat integrated safety glasses
Proveedor: 3M
MMMAV9CEA 21.9 EUR
MMMAV9C
Hard hat integrated safety glasses
Gafas
Hard hat integrated safety glasses, V9C, are clear lens safety glasses designed to attach to 3M hard hats for protection from particles and UV.
  • Provides eye protection from particles and UV
  • Easily attaches to 3M hard hats
  • Durable anti-scratch and anti-fog lens coating helps minimise physical damage and fogging
  • Easy to adjust for a good fit
  • Faceshield marking 2C-1,2 3M 1 F
Hard hat integrated safety glasses, V9C, feature a unique design that allows them to slide easily between the hard hat shell and headband, fitting closely to the wearer’s cheeks to provide protection.
Certificaciones: EN 166:2001