Label printers with Brady workstation LAB suite, BMP™ 51
Proveedor: Brady
BRDY306856EA 932 EUR
116-0950 BRDY306856
Label printers with Brady workstation LAB suite, BMP™ 51
Impresoras Label Printers
The BMP™ 51 Label Maker is a compact, versatile portable printer with built-in Bluetooth® technology for wireless printing.
  • 300 dpi
  • Mono colour
  • Applications include cable and wire identification, datacom and audio, visual equipment labelling, electrical identification, general and industrial identification, laboratory identification, and panel identification
This label maker has a 300 dpi print head and can print professional, durable labels up to 38.10 mm wide. It prints on continuous and die-cut materials at a speed of 25.40 mm per second. With a built-in cutter, easy top-loading cartridges, and 3 different options for battery power, the BMP™51 Label Maker is a powerful printer for on-the-job label making. The BMP™51 has a full QWERTY keyboard, for stand alone operation. The BMP™51 Label Maker is ideal for electrical, datacom, facility/safety, product and panel manufacturing, and laboratory labelling applications. The BMP™51 comes with the Brady Workstation Product & Wire Identification Software suite enabling you to immediately start designing professional labels. This suite combines frequently used software apps to create product & cable identification labels.
Materials supported include Freezerbondz Polyester; Heat-shrink Polyolefin Sleeve; Metallised Polyester; Nylon Cloth; Polyester; Polyethylene Tag; Polypropylene; Polypropylene Tag; Reflective Tape; Self-laminating Polyester; Self-laminating Vinyl; StainerBondz Polyester; Tamper-resistant Vinyl; Vinyl; Vinyl Cloth. Barcode type Code 39 and 128, other via software. Manual cut; Between labels or at end of print job; With label retention.
Languages: Danish; Dutch; English; Finnish; French; German; Italian; Norwegian (Bokmål); Portuguese; Spanish; Swedish.
Información de suministro: This printer ships in a hard case with a Rechargeable Lithium Ion Battery, USB Cable, AC adaptor + EU or UK power cord, Product CD, Brady Printer Drivers CD, Quick Start Guide, a Bluetooth network card, a Brady Workstation Laboratory Identification Suite and an MC1-1000-422 cartridge.