6-Quinolineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Proveedor: TCI EUROPE

TCIAT3507-1GEA 56 EUR
TCIAT3507-1G
6-Quinolineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
6-Quinolineboronic acid pinacol ester
Formula: C₁₅H₁₈BNO₂
MW: 255,1199 g/mol
Número MDL: MFCD06411327
Núm. CAS: 406463-06-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 67.0 to 71.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR