3-Quinolineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Proveedor: TCI EUROPE
Atención

TCIAT3180-1GEA 69 EUR
TCIAT3180-1G
3-Quinolineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3-Quinolineboronic acid pinacol ester
Formula: C₁₅H₁₈BNO₂
MW: 255,1199 g/mol
Número MDL: MFCD05155218
Núm. CAS: 171364-85-5

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 58.0 to 62.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR