meso-Tetrafenilporfina ≥98.0% (by HPLC) chlorin-free
Proveedor: TCI EUROPE
Sinónimos:
5,10,15,20-Tetrafenil-21H,23H-porfina
TCIAT1359-1GEA
139
EUR
TCIAT1359-1G
meso-Tetrafenilporfina ≥98.0% (by HPLC) chlorin-free
meso-Tetrafenilporfina
Formula:
C₄₄H₃₀N₄ MW: 614,75 g/mol |
Número MDL:
MFCD00011680 Núm. CAS: 917-23-7 EINECS: 213-025-9 |
Resultados test especificaciones
Appearance | Orange to Brown to Dark purple powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Tetraphenylchlorin | max. 0.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...