Phloxin B ≥85.0% (by UV/Vis)
Proveedor: TCI EUROPE
Sinónimos:
CI 45410
TCIAT0050-25GEA
50
EUR
TCIAT0050-25G
Phloxin B ≥85.0% (by UV/Vis)
Phloxin B
Formula:
C₂₀H₂Br₄Cl₄Na₂O₅ MW: 829,6344 g/mol |
Número MDL:
MFCD00070627 EINECS: 242-355-6 |
Resultados test especificaciones
Appearance | Dark red to Brown powder to crystal |
Purity (UV-vis method) | min. 85.0 % (calcd.on dried substance) |
Lambda max. | 536.0 to 540.0 nm (0.02mol/L AcONH4 sol.) |
Drying loss | max. 7.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...