Disialylnonasaccharide-beta-pNP ≥98.0% (by HPLC)

Proveedor: TCI EUROPE

TCIAN0913-1MGEA 198 EUR
TCIAN0913-1MG
Disialylnonasaccharide-beta-pNP ≥98.0% (by HPLC)
Disialylnonasaccharide-beta-pNP
Formula: C₉₀H₁₄₁N₇O₆₄
MW: 2345,10 g/mol
Temperatura de almacenaje: Congelador
Núm. CAS: 1408055-26-4

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR