4-Nitrosodiphenylamine ≥97.0% (by HPLC)

Proveedor: TCI EUROPE
Atención

TCIAN0263-25GEA 214 EUR
TCIAN0263-25G
4-Nitrosodiphenylamine ≥97.0% (by HPLC)
4-Nitrosodiphenylamine
Formula: C₁₂H₁₀N₂O
Número MDL: MFCD00002062
Núm. CAS: 156-10-5
EINECS: 205-848-7

Order Now

Resultados test especificaciones

Appearance Red to Dark blue to Black powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 140.0 to 145.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR