Ginkgolide B ≥93.0% (by HPLC)
Proveedor: TCI EUROPE
TCIAG0553-25MGEA
67
EUR
TCIAG0553-25MG
TCIAG0553-100MG
Ginkgolide B ≥93.0% (by HPLC)
Ginkgolide B
Formula:
C₂₀H₂₄O₁₀ MW: 424,40 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD05662347 Núm. CAS: 15291-77-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 93.0 area% |
Elemental analysis (Carbon) | 54.00 to 59.00 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...