3-Fluoro-4-iodobenzonitrile ≥98.0% (por GC)
Proveedor: TCI EUROPE
|
Peligro
|
TCIAF1093-1GEA
62
EUR
TCIAF1093-1G
3-Fluoro-4-iodobenzonitrile ≥98.0% (por GC)
3-Fluoro-4-iodobenzonitrile
Formula:
C₇H₃FIN MW: 247,01 g/mol |
Número MDL:
MFCD07782076 Núm. CAS: 887266-99-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 113.0 to 117.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...