3-Fluoro-2-methylphenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI EUROPE
|
Atención
|
TCIAF0955-1GEA
105
EUR
TCIAF0955-1G
3-Fluoro-2-methylphenylboronic acid (contains varying amounts of Anhydride)
3-Fluoro-2-methylphenylboronic acid
Formula:
C₇H₈BFO₂ MW: 153,95 g/mol |
Número MDL:
MFCD07368679 Núm. CAS: 163517-61-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 114.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...