2-Ethyl-3-hydroxy-6-methylpyridine ≥98.0% (by GC, titration analysis)
Proveedor: TCI EUROPE
|
Atención
|
TCIAE0833-5GEA
81
EUR
TCIAE0833-5G
2-Ethyl-3-hydroxy-6-methylpyridine ≥98.0% (by GC, titration analysis)
2-Ethyl-3-hydroxy-6-methylpyridine
Formula:
C₈H₁₁NO MW: 137,1800 g/mol |
Número MDL:
MFCD00462409 Núm. CAS: 2364-75-2 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 167.0 to 171.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...