Calcium disodium EDTA hydrate ≥98.0% (by titrimetric analysis)

TCIAE0088-25GEA 22 EUR
TCIAE0088-25G
Calcium disodium EDTA hydrate ≥98.0% (by titrimetric analysis)
Calcium disodium EDTA hydrate
Número MDL: MFCD00149453
Núm. CAS: 23411-34-9
EINECS: 200-529-9

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water 8.0 to 13.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR