3,4-Diaminobenzonitrile ≥98.0% (por GC)
Proveedor: TCI EUROPE
|
Peligro
|
TCIAD5194-5GEA
125
EUR
TCIAD5194-5G
TCIAD5194-1G
3,4-Diaminobenzonitrile ≥98.0% (por GC)
3,4-Diaminobenzonitrile
Formula:
C₇H₇N₃ MW: 133,15 |
Número MDL:
MFCD00723901 Núm. CAS: 17626-40-3 UN: 3439 ADR: 6.1,III |
Resultados test especificaciones
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 147.0 to 151.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...