2,4-Dibromo-6-[(E)-[[(1R,2R)-2-(isoindolin-2-yl)-1,2-diphenylethyl]imino]methyl]phenol ≥93.0% (by HPLC)

Proveedor: TCI EUROPE

TCIAD5160-50MGEA 233 EUR
TCIAD5160-50MG
2,4-Dibromo-6-[(E)-[[(1R,2R)-2-(isoindolin-2-yl)-1,2-diphenylethyl]imino]methyl]phenol ≥93.0% (by HPLC)
2,4-Dibromo-6-[(E)-[[(1R,2R)-2-(isoindolin-2-yl)-1,2-diphenylethyl]imino]methyl]phenol
Formula: C₂₉H₂₄Br₂N₂O
MW: 576,33 g/mol
Temperatura de almacenaje: Congelador
Núm. CAS: 1420042-07-4

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 93.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR