Resodiacetophenone ≥98.0% (by GC, titration analysis)

Proveedor: TCI EUROPE
Atención

TCIAD5102-5GEA 49 EUR
TCIAD5102-5G
Resodiacetophenone ≥98.0% (by GC, titration analysis)
Resodiacetophenone
Formula: C₁₀H₁₀O₄
MW: 194,1840 g/mol
Número MDL: MFCD00017655
Núm. CAS: 2161-85-5
EINECS: 218-482-8

Order Now

Resultados test especificaciones

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 181.0 to 185.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR