(1S,3R)-(-)-Camphoric acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI EUROPE
|
Atención
|
TCIAC3237-1GEA
102
EUR
TCIAC3237-1G
(1S,3R)-(-)-Camphoric acid ≥98.0% (by GC, titration analysis)
(1S,3R)-(-)-Camphoric acid
Formula:
C₁₀H₁₆O₄ MW: 200,23 g/mol |
Número MDL:
MFCD00064937 Núm. CAS: 560-09-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 186.0 to 190.0 deg-C |
Specific rotation [a]20/D | -46.0 to -50.0 deg (C=4,EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...