3-Chloro-2-hydroxy-5-nitropyridine ≥98.0% (by titrimetric analysis)

Proveedor: TCI EUROPE
Atención

TCIAC3185-1GEA 91 EUR
TCIAC3185-1G
3-Chloro-2-hydroxy-5-nitropyridine ≥98.0% (by titrimetric analysis)
3-Chloro-2-hydroxy-5-nitropyridine
Formula: C5H3ClN2O3
MW: 174,5400 g/mol
Número MDL: MFCD17015099
Núm. CAS: 22353-38-4

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 195.0 to 199.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR