5-Chloropyrazine-2-carboxylic acid ≥98.0% (by titrimetric analysis)
Proveedor: TCI EUROPE
TCIAC3002-1GEA
64
EUR
TCIAC3002-1G
5-Chloropyrazine-2-carboxylic acid ≥98.0% (by titrimetric analysis)
5-Chloropyrazine-2-carboxylic acid
Formula:
C5H3ClN2O2 MW: 158,54 g/mol |
Número MDL:
MFCD09033269 Núm. CAS: 36070-80-1 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...