tert-Butyl (S)-2-(2,6-Dichlorophenyl)-5-oxo-3-[(S)-1-phenylethyl]imidazolidine-1-carboxylate ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI EUROPE

TCIAB6109-1GEA 300 EUR
TCIAB6109-1G
tert-Butyl (S)-2-(2,6-Dichlorophenyl)-5-oxo-3-[(S)-1-phenylethyl]imidazolidine-1-carboxylate ≥98.0% (by HPLC, total nitrogen)
tert-Butyl (S)-2-(2,6-Dichlorophenyl)-5-oxo-3-[(S)-1-phenylethyl]imidazolidine-1-carboxylate
Formula: C₂₂H₂₄Cl₂N₂O₃
MW: 435,35 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD30187601
Núm. CAS: 1149765-08-1

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Optical purity (LC) min. 98.0 ee%
Melting point 99.0 to 103.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR