[2,2'-Binaphthalen]-6-ylboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI EUROPE
Atención

TCIAB5055-1GEA 201 EUR
TCIAB5055-1G
[2,2'-Binaphthalen]-6-ylboronic acid (contains varying amounts of Anhydride)
[2,2'-Binaphthalen]-6-ylboronic acid
Formula: C₂₀H₁₅BO₂
MW: 298,15 g/mol
Núm. CAS: 1025456-44-3

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 and le 106.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR