5-Bromo-8-nitroisoquinoline ≥98.0% (by GC, titration analysis)
Proveedor: TCI EUROPE
|
Atención
|
TCIAB4163-1GEA
57
EUR
TCIAB4163-1G
5-Bromo-8-nitroisoquinoline ≥98.0% (by GC, titration analysis)
5-Bromo-8-nitroisoquinoline
Formula:
C₉H₅BrN₂O₂ MW: 253,0500 g/mol |
Número MDL:
MFCD02091227 Núm. CAS: 63927-23-1 |
Resultados test especificaciones
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 138.0 to 142.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...