7-Aminoquinoline ≥98.0% (by GC, titration analysis)

Proveedor: TCI EUROPE
Atención

TCIAA3268-1GEA 105 EUR
TCIAA3268-1G
7-Aminoquinoline ≥98.0% (by GC, titration analysis)
7-Aminoquinoline
Formula: C₉H₈N₂
MW: 144,18 g/mol
Número MDL: MFCD01646243
Núm. CAS: 580-19-8

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water max. 7.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR