2-Amino-4-(4-bromophenyl)thiazole ≥98.0% (by GC, titration analysis)

Proveedor: TCI EUROPE
Atención

TCIAA2829-5GEA 63 EUR
TCIAA2829-5G
2-Amino-4-(4-bromophenyl)thiazole ≥98.0% (by GC, titration analysis)
2-Amino-4-(4-bromophenyl)thiazole
Formula: C₉H₇BrN₂S
Número MDL: MFCD00051953
Núm. CAS: 2103-94-8

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 184.0 to 188.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR