(R)-2-(Aminomethyl)-1-N-boc-pyrrolidine ≥98.0% (by titrimetric analysis)
Proveedor: TCI EUROPE
|
Peligro
|
TCIAA2712-1GEA
211
EUR
TCIAA2712-1G
(R)-2-(Aminomethyl)-1-N-boc-pyrrolidine ≥98.0% (by titrimetric analysis)
(R)-2-(Aminomethyl)-1-N-boc-pyrrolidine
Formula:
C₁₀H₂₀N₂O₂ MW: 200,28 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD03419256 Núm. CAS: 259537-92-3 UN: 2735 ADR: 8,III |
Resultados test especificaciones
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +46.0 to +50.0 deg (C=1、CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...