(S)-1-Boc-2-(aminomethyl)pyrrolidine ≥98.0% (by GC, titration analysis)
Proveedor: TCI EUROPE
|
Peligro
|
TCIAA2473-1GEA
113
EUR
TCIAA2473-1G
(S)-1-Boc-2-(aminomethyl)pyrrolidine ≥98.0% (by GC, titration analysis)
(S)-1-Boc-2-(aminomethyl)pyrrolidine
Formula:
C₁₀H₂₀N₂O₂ MW: 200,28 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD03419257 Núm. CAS: 119020-01-8 UN: 2735 ADR: 8,III |
Resultados test especificaciones
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -46.0 to -50.0 deg (C=1、,CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...