N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulphonamide ≥98.0% (by titrimetric analysis)

Proveedor: TCI EUROPE
Atención

TCIAA1548-1GEA 78 EUR
TCIAA1548-1G
N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulphonamide ≥98.0% (by titrimetric analysis)
N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulphonamide
Formula: C₂₁H₂₂N₂O₂S
MW: 366,48 g/mol
Número MDL: MFCD02093428
Núm. CAS: 144222-34-4

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 128.0 to 131.0 deg-C
Specific rotation [a]20/D -25.0 to -35.0 deg (C=1, CH3CN)
Solubility in hot Acetonitrile almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR