MERC105235
1.05235.0025EA 388 EUR
1.05235.0025
MERC105235
Certistain® dry dye used to stain nuclei and Nissl bodies
The CERTISTAIN® dry dye Cresyl violet (acetate) is used for the preparation of a staining solution that can be used for the staining of nuclei and Nissl bodies. Further, it is also applied in the standard stain for the identification of Helicobacter and sex chromatin (X chromatin).
As CERTISTAIN® dye it is chemically quality-certified according to highest standards and strict specifications. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations.