Muffle furnaces with fold down door, Tmax 1300 °C and standard controller B510, L 5/13 - L 15/13
Proveedor: Nabertherm
Nabertherm
455-1569EA 7430 EUR
455-1569 455-1349 455-1562 455-1561 455-1146 455-1145 455-1563 455-1148 455-1357 455-1356 455-1568 455-1139 455-1359 455-1138 455-1140 455-1571 455-1351 455-1350
Muffle furnaces with fold down door, Tmax 1300 °C and standard controller B510, L 5/13 - L 15/13
Hornos Hornos
These furnaces feature heating elements on supporting tubes that radiate freely into the chamber, ensuring particularly short heating times. With the robust lightweight refractory brick insulation, they can reach a max. working temperature of 1300 °C.
  • Heating from two sides
  • Multi-layer insulation with robust lightweight refractory bricks in the furnace chamber
  • Textured stainless steel housing
  • Dual shell housing for low external temperatures and high stability
  • Fold down door (L models) which can be used as a work platform
  • Exhaust air outlet on rear wall of furnace
The furnace is equipped with the B510 standard controller with touch screen display, featuring 5 freely adjustable programs and 4 segments and 2 extra functions. These controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation. The B510 controllers can automatically activate the vent with fan or catalyst (not model L 15).
Información para pedidos: All units require a 230 V single-phase power supply. Optional accessories e.g. a vent, vent with fan or catalytic converter, over-temperature limit controller with adjustable temperature cut out for thermal protection Class 2, in accordance with EN 60519-2 as temperature limiter to protect the oven and load, protective gas connection on the rear wall of the furnace. Manual or automatic gas supply systems are also available. Corresponding models with lift up doors, different Tmax and other controllers are also available. Please search the website for more details.
Precaución: Maximum continuous operating temperature should be 50 °C below the 'Max. temp.' of the model.
Note heat up times: If connected at 230 V 1/N/PE rsp. 400 V 3/N/PE.
*Temperature uniformity: ± 5 K with closed air inlet in the empty workspace according to DIN 17052-1 at working temperatures > 800 °C