Muffle furnaces with scale and software, Tmax 1100 °C, L 9/11 SW or LT 9/11 SW
Proveedor: Nabertherm
Nabertherm
NABELW092H1TN1ZEA 10590 EUR
NABELW092H1TN1Z NABELW092K1RN1Z NABELW092K1TN1Z NABELW092H1RN1T NABELW092H1RN1Z NABELW094K1ON1 455-2035 455-2056 455-2034 455-2036 NABELW092H1RN1TZ 455-2038 NABELW092K1TN1 NABELW092K1TN1T NABELW092K1RN1 NABELW092H1TN1T 455-2053 NABELW092K1RN1TZ 455-2052 455-2030 455-2054 NABELW092K1RN1T 455-2032 NABELW094K1ON1TZ NABELW092K1TN1TZ NABELW094K1ON1Z NABELW094K1ON1T NABELW092H1RN1 NABELW092H1TN1 455-2028 NABELW092H1TN1TZ 455-2029
Muffle furnaces with scale and software, Tmax 1100 °C, L 9/11 SW or LT 9/11 SW
Hornos Hornos
These weighing furnaces with an integrated precision scale and software, were designed especially for determining combustion loss in the laboratory. The determination of combustion loss is necessary, for instance, when analysing sludges. Combustion loss is defined as the difference between the initial total mass and the combustion residue. During the process the software supplied records both temperature and weight loss.
  • Heating from two sides
  • Ceramic heating plates with integral heating element which is protected from fumes and splashing, and easy to replace
  • Only fibre materials which are not classified as carcinogenic according to TRGS 905, class 1 or 2 are used
  • Optional fold down door (L) which can be used as a work platform or lift up door (LT) with hot surface facing away from the operator
  • Process control and documentation for temperature and combustion loss via VCD software package for monitoring, documentation and control
These furnaces are equipped either with the B510 standard controller with touch screen display, featuring 5 freely adjustable programs and 4 segments and 2 extra functions or the optional C550 controller with touch screen display, featuring 10 freely adjustable programs and 20 segments and 2 extra functions.
These touch screen controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation.
Minutes to Tmax without load and standard voltage: 75
Información para pedidos: Optional accessories such as a vent, vent with fan or catalytic converter to influence the weighing results, over-temperature limit controller with adjustable temperature cut out for thermal protection Class 2, in accordance with EN 60519-2 as temperature limiter to protect the oven and load, access port for thermocouple. Corresponding models with different Tmax are also available. Please search the website for more details.
Información de suministro: Delivery includes base, ceramic plunger with base plate in the furnace lining, precision scale - four scales available for different maximum weights and scaling ranges and software package. The included standard scale is up to a weight range of 2200 g (less 850 g for the plunger) and has a readability of 0,01 g with a minimum load of 0,5 g. There are also optional scales with a weight range of 4200, 6200 or 12000 g available, please consult your Avantor representative for further information.
Precaución: Maximum continuous operating temperature should be 50 °C below the 'Max. temp.' of the model shown in the table above.
Outer dimensions include opened lift up door for LT models.